VWR® Micro Star 21/21R Microcentrifuges, Ventilated/Refrigerated
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
521-2656EA 17390 DKK
521-2656 521-2657
VWR® Micro Star 21/21R Microcentrifuges, Ventilated/Refrigerated
Centrifuger Mikrocentrifuger
Micro Star 21 and 21R microcentrifuges combine power, versatility and convenience in a safe, compact, easy to use laboratory instrument. Both the ventilated Micro Star 21 and the refrigerated Micro Star 21R are excellent choice for routine sample preparation process and for high-end research applications. The special biocontainment rotor lid, for one-click rotor opening and closing, shortens retrieval time compared to screw on lids.
  • Quiet and powerful performance up to 21100×g
  • ClickSeal® biocontainment rotor lid keeps the user and samples safe
  • Easy