n-Heptan ≥99.0% (by GC), Purum

Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: high purity solvents

51750-25L
51750-25LEA 9040 DKK
51750-25L
n-Heptan ≥99.0% (by GC), Purum
n-Heptane
Formel: H₃C(CH₂)₅CH₃
Molekylvægt: 100.20 g/mol
Kogepunkt: 98.2…98.4 °C (1,000 hPa)
Smeltepunkt: –90.5 °C
Massefylde: 0.69 g/cm3 (15 °C)
Flammepunkt: –4 °C
MDL Number: MFCD00009544
CAS-nummer: 142-82-5
UN nummer: 1206
ADR: 3 II
Merck Index: 13,04679

Bestil nu

Specifikationer Testresultater

Appearance (color) complying
Appearance (form) complying
IR spectrum complying
Assay (GC) Min. 99.0 %
Density (D 20/4) 0.682 - 0.686
Refractive index (n 20/D) 1.387 - 1.389
Water (Karl Fischer) Max. 0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR