Ultrafiltration devices, Centrifree®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

4104
516-6570EA 4170 DKK
516-6570
Ultrafiltration devices, Centrifree®
Filtre Centrifugefiltre
The Centrifree® Ultrafiltration device with Ultracel® regenerated cellulose membrane is ideal for separating free from bound microsolutes in serum, plasma, and other biological samples. The device is licensed for research use and in vitro diagnostic use. It holds a sample volume of 1 ml with a hold-up volume of 10 µl.

  • Separate unbound (free) therapeutic drugs, testosterone, thyroxin, etc.
  • Enables accuracy in binding studies
  • Critical for new drug investigations
  • Facilitates deproteinisation to reduce complexity or matrix interference of biological samples
More Product Information
Bestil nu

Specifikationer

Filtreringsareal 0,92 cm²
Environmentally Preferable
Dødvolumen 10 µl
Ø×H 16×95 mm
Prøvevolumen 1000 µl

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR