Sprøjteløse filtre, UniPrep™, Whatman™
Leverandør: Whatman products (Cytiva)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
UN113UGMF UN113UNYL UN113UORG UN513UAQU
514-0574EA 3120 DKK
514-0574 514-0129 514-0130 516-0675
Sprøjteløse filtre, UniPrep™, Whatman™
Filtre Filtre uden sprøjter
UniPrep™ Syringeless Filters are preassembled filtration devices for the filtration and storage of laboratory samples. These devices are quick and easy to use and feature a plunger, filter, and vial in one unit. They replace syringe-coupled filtration devices with single, disposable units.
  • Integral storage vial saves time and minimizes laboratory waste
  • Built-in glass fiber prefilter means even difficult samples are quick and easy to prepare
  • Choice of membranes for wide sample compatibility
  • Recyclable after use
UniPrep™ devices consist of two parts: aMore Product Information