Vakuumfiltreringssystemer på flasketop, PES-membran, Rapid-Flow™ Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
564-0020
514-0590EA 1460 DKK
514-0590
Vakuumfiltreringssystemer på flasketop, PES-membran, Rapid-Flow™ Nalgene®
Filtre Topfiltre til flasker
Rapid-Flow™ disposable filter units with PES membrane, PS funnel and PP collection tube feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination and optimal choice for small volume tissue culture work. Offers clean, fast filtration of 20 to 50 ml of media or buffers.
  • Color coded according to membrane
  • Fastest flow rate and lowest clogging of comparable alternatives
  • 0.1 µm PES available for mycoplasma protection
  • 0.2 µm PES devices are stem cell and