Bottle top vacuum filtration systems, cellulose nitrate membrane, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
140-4045
513-1402EA 1050 DKK
513-1402
Bottle top vacuum filtration systems, cellulose nitrate membrane, Nalgene®
Filtre Topfiltre til flasker
These filters are particularly useful for large volume microbiological QC samples.
  • Convenient snap-away design
  • Compatible with filter manifolds and filtering flasks
Certificeringer: Membrane conforms to APHA standard methods, 16th edition, section 909A, B, C. Meets EPA requirements for water quality work.
Leveringsinformation: Upper chamber with reference mark at 150 ml; PS cover; CN membrane; support pad; PS support plate/funnel that fits a stopper or adapter with ⁹/₁₆" hole; quick-disconnect adapter for ¹/₄ or ⁵/₁₆" tubing. Each pack of 12More Product Information
Bestil nu
Specifikationer
Environmentally Preferable Lav fremstillingspåvirkning
Miljøcertificering ISO14001 (Certificate Registration No. EMS 752603)
NAR Certificate of Retirement