Filter plates, 96-well, for solvent filtration, AcroPrep™ Advance
Leverandør: Cytiva (Formerly Pall Lab)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8047 8048 8247 8148 8147 8248 8582 8584 8682 8684 8782 8784 8586 8588 8686 8688
738-0109EA 2220 DKK
738-0109 738-0110 738-0131 738-0122 738-0121 738-0132 513-0163 513-0164 514-1096 514-1097 514-1098 514-1099 513-0166 513-0168 513-0167 513-0169
Filter plates, 96-well, for solvent filtration, AcroPrep™ Advance
Mikroplader Filterplader
The AcroPrep™ Advance filter plate portfolio includes a variety of membrane types and pore sizes that provide the best possible performance for a wide range of sample preparation and high throughput screening applications. Available with a choice of PTFE or PTFE membrane.
  • Kemiske resistente materialer leverer en stabil platform til forarbejdning af prøver i organiske opløsningsmidler
  • Lav uspecifik binding - inerte konstruktionsmaterialer sikrer fuldkommen prøvegenindvinding
  • Brøndgeometrien resulterer i hurtigere og mere ensartede filtreringsrater på tværs