VWR®, Büchner funnels
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
511-0049EA 345 DKK
511-0049 511-0069 511-0068 511-0067 511-0066 511-0065 511-0059 511-0058 511-0057 511-0055 511-0048 511-0075 511-0056
VWR®, Büchner funnels
Tragte Filtreringstragte Büchner-tragte
White glazed porcelain.
For paper or membrane filtration discs.