Methylenblåt solution for microbiology, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Basic Blue 9, Methylthioninium chloride, Methylene blue chloride, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride, Swiss Blue

50484-100ML-F
50484-100ML-FEA 246 DKK
50484-100ML-F
Methylenblåt solution for microbiology, Sigma-Aldrich®
Methylenblåt

Ehrlich’s reagent III, Loeffler’s Methylene Blue. Methylene Blue was earlier recognized as a good nuclear stain, the staining solutions are always either aged or alkalized, if not both. More importantly it is an excellent stain for ribonucleic acid, which is the important stainable constituent of bacterial bodies, of lymphocyte and hemoportoal cytoplasm, and of nerve cell tigroid granules. It can also be employed in capsule stain. The capsule stain begins as a negative stain leaving the organisms colorless. The capsule are clearly seen as halo surrounding the bacterial cell. Another staining procedure is performed with nigrosin (Cat. No. 43925).

Formel: C₁₆H₁₈ClN₃S
MDL Number: MFCD00150008
CAS-nummer: 61-73-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b393708522bf6442a679105dac615d265bafa5a&vid=5b393708522bf6442a679105dac615d265bafa5a&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR