Acetone, Finyte® Microelectronic, J.T.Baker®

Leverandør: Avantor
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Propanon

5018-20EA 0 DKK
5018-20
Acetone, Finyte® Microelectronic, J.T.Baker®
Acetone
Formel: CH₃COCH₃
Molekylvægt: 58.08 g/mol
Kogepunkt: 56.2
Smeltepunkt: -95.4
MDL Number: MFCD00008765
CAS-nummer: 67-64-1
UN nummer: 1090
ADR: 3 II
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

For Microelectronic Use = 99.5 %
Assay (by GC) ≥ 99.5 %
Color (APHA) ≤ 10
Acidity (µeq/g) ≤ 0.3
Aldehydes (as HCHO) ≤ 20000 ppb
Alkalinity (µeq/g) ≤ 0.5
Isopropyl Alcohol (CH₃CHOHCH₃)(by GC) ≤ 0.05 %
Methanol (CH₃OH)(by GC) ≤ 0.05 %
Sulfate (SO₄), For Information Only = 3000.0 ppb
Residue after Evaporation ≤ 3000.0 ppb
Water (H₂O) ≤ 0.50 %
Solubility in H₂O Passes Test
Chloride (Cl) ≤ 200.0 ppb
Phosphate (PO₄) ≤ 50.0 ppb
Trace Impurities - Aluminum (Al) ≤ 10.0 ppb
Trace Impurities - Antimony (Sb) ≤ 5.0 ppb
Trace Impurities - Arsenic (As) ≤ 5.0 ppb
Trace Impurities - Barium (Ba) ≤ 10.0 ppb
Trace Impurities - Beryllium (Be) ≤ 10.0 ppb
Trace Impurities - Bismuth (Bi) ≤ 10.0 ppb
Trace Impurities - Boron (B) ≤ 10.0 ppb
Trace Impurities - Cadmium (Cd) ≤ 10.0 ppb
Trace Impurities - Calcium (Ca) ≤ 10.0 ppb
Trace Impurities - Chromium (Cr) ≤ 10.0 ppb
Trace Impurities - Cobalt (Co) ≤ 10.0 ppb
Trace Impurities - Copper (Cu) ≤ 10.0 ppb
Trace Impurities - Gallium (Ga) ≤ 10.0 ppb
Trace Impurities - Germanium (Ge) ≤ 10.0 ppb
Trace Impurities - Gold (Au) ≤ 10.0 ppb
Trace Impurities - Iron (Fe) ≤ 10.0 ppb
Trace Impurities - Lead (Pb) ≤ 10.0 ppb
Trace Impurities - Lithium (Li) ≤ 10.0 ppb
Trace Impurities - Magnesium (Mg) ≤ 10.0 ppb
Trace Impurities - Manganese (Mn) ≤ 10.0 ppb
Trace Impurities - Molybdenum (Mo) ≤ 10.0 ppb
Trace Impurities - Nickel (Ni) ≤ 10.0 ppb
Trace Impurities - Niobium (Nb) ≤ 10.0 ppb
Trace Impurities - Potassium (K) ≤ 10.0 ppb
Trace Impurities - Silicon ≤ 10.0 ppb
Trace Impurities - Silver (Ag) ≤ 10.0 ppb
Trace Impurities - Sodium (Na) ≤ 10.0 ppb
Trace Impurities - Strontium (Sr) ≤ 10.0 ppb
Trace Impurities - Tantalum (Ta) ≤ 10.0 ppb
Trace Impurities - Thallium (Tl) ≤ 10.0 ppb
Trace Impurities - Tin (Sn) ≤ 10.0 ppb
Trace Impurities - Titanium (Ti) ≤ 10.0 ppb
Trace Impurities - Vanadium (V) ≤ 10.0 ppb
Trace Impurities - Zinc (Zn) ≤ 10.0 ppb
Trace Impurities - Zirconium (Zr) ≤ 10.0 ppb
Particle Count at point of fill - 0.5 µm and greater ≤ 150 par/ml
Particle Count at point of fill - 1.0 µm and greater ≤ 25 par/ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR