Ammoniumacetat 98 Alkalimetric, LiChropur™ LC-MS, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Acetic acid ammonium salt

MQ100

5.33004.0050
5.33004.0050EA 1197 DKK
5.33004.0050
Ammoniumacetat 98 Alkalimetric, LiChropur™ LC-MS, Supelco®
Ammoniumacetat
Formel: H₃CCOONH₄
Molekylvægt: 77.08 g/mol
Smeltepunkt: 114 °C
Massefylde: 1.17 g/cm³ (20 °C)
Flammepunkt: 136
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00013066
CAS-nummer: 631-61-8
EINECS: 211-162-9
Merck Index: 13,00495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR