Supelco® Organic Reference Standard, (±)-Geosmin and 2-Methylisoborneol Solution

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

4M7525-U
4M7525-UEA 4030 DKK
4M7525-U
Supelco® Organic Reference Standard, (±)-Geosmin and 2-Methylisoborneol Solution
Standarder Økologiske standarder

  • GHS signal: GHS02, GHS06, GHS08


Advarsel: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311

Bestil nu

Specifikationer

Environmentally Preferable
Fareklassebeskrivelse H225 - H301 + H311 + H331 - H370
Shipping conditions STD
Storage conditions RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR