Nordihydroguaiaretic acid, Millipore®

Leverandør: Merck Millipore Calbiochem
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

479975-250
479975-250EA 1020 DKK
479975-250
Nordihydroguaiaretic acid, Millipore®
Nordihydroguaiaretic acid
Formel: C₁₈H₂₂O₄
Molekylvægt: 302.37 g/mol
MDL Number: MFCD00002206
CAS-nummer: 500-38-9
EINECS: 207-903-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR