Menadione (2-methyl-1,4-naphthoquinone, vitamin K3)

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Menadione, 1,4-Dihydro-2-methylnaphthalene-1,4-dione, 2-Methylnaphthalene-1,4-dione

47775
47775.EA 495 DKK
47775.
Menadione (2-methyl-1,4-naphthoquinone, vitamin K3)
Menadione (2-methyl-1,4-naphthoquinone, vitamin K3)
Formel: C₁₁H₈O₂
Molekylvægt: 172.19 g/mol
MDL Number: MFCD00001681
CAS-nummer: 58-27-5
EINECS: 200-372-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4abf21c8c7b87ecc5d76f06c49fe58f257df124&vid=0cc46e2c7abff807e6e91dbb65784b9b0dce6ea8&prid=web&clid=SAPDAM

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR