3,3'-Dihydroxybenzidine 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4,4'-Diamino-3,3'-dihydroxybiphenyl, 3,3'-Dihydroxy-4,4'-diaminobiphenyl, 4,4'-Diamino-[1,1'-biphenyl]-3,3'-diol, Benzidine-3,3'-diol

474450010 474450050 474450250
474450010.EA 323 DKK
474450010. 474450050. 474450250.
3,3'-Dihydroxybenzidine 98%
3,3'-Dihydroxybenzidine
Formel: C₁₂H₁₂N₂O₂
Molekylvægt: 216.24 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00039149
CAS-nummer: 2373-98-0
EINECS: 220-190-0
New Product

Bestil nu

Specifikationer Testresultater

Appearance white to brown powder or crystals
Infrared spectrum Conforms
HPLC ≥97.5 %

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR