Pharmacy refrigerators, MKv and MKUv to DIN 58345
Leverandør: Liebherr
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MKUV1610 MKUV1613
472-0121EA 15749 DKK
472-0121 471-1053
Pharmacy refrigerators, MKv and MKUv to DIN 58345
Køleskabe
These pharmacy refrigerators are specifically designed for the safe storage of pharmacy products compliant with DIN 58345. They provide maximum storage space on a 60 cm footprint and the under bench models are only 820 mm high (MKUv) so will fit under almost any bench.
  • Electronic temperature control system with 0,1 °C temperature adjustment
  • Audible and visual alarms for door open and temperature (user-adjustable)
  • RS485 interface and volt-free contact for central monitoring
  • Interchangeable self-closing door with lock
  • LED interior ceiling light, with switch