Supelco® Organic Reference Standard, Corn oil

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

47112-U
47112-UEA 511 DKK
47112-U
Supelco® Organic Reference Standard, Corn oil
Standarder Økologiske standarder

Bestil nu

Specifikationer

Synonym(s) Maize oil
Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 8001-30-7
MDL No. MFCD00130865
Flammepunkt 254 °C
Shipping conditions STD
Storage conditions 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR