Professional ventilated refrigerators, GKPv
Leverandør: Liebherr
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
GKPV6520 GKPV6590 GKPV1490 GKPV6573
472-1063EA 21210 DKK
472-1063 471-0910 471-0909 471-1066
Professional ventilated refrigerators, GKPv
Køleskabe
These commercial refrigerators feature a fan for forced air cooling with temperature adjustable from –2 to +15 °C or +1 to +15 °C.
  • Electronic temperature regulation, with digital display
  • Alarm for temperature and door opening
  • Automatic defrosting with evaporation of condensates
  • Interior with rounded corners to make cleaning easier
  • Door is lockable and reversible, self closing at 45° but stable at 90°
Interior and exterior made from stainless steel AISI 306, with indirect ventilated cooling, insulation 83 mm (allows temperature controlMore Product Information