Universalskabsfrysere, G modeller
Leverandør: Liebherr
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
G2713 G2413 G2033 G2433 G2733
471-0240EA 0 DKK
471-0240 472-2339 471-1020 471-0911 471-0912
Universalskabsfrysere, G modeller
Frysere Laboratorie-/frysere til almindelige formål
These are general purpose freezers with static cooling - ideal for the storage of non critical products. Spacious, energy-saving, these upright freezers are very economical with optimised cooling systems and compressors. They use very little electricity and are very user friendly. All models are equipped with a recovery defrost water system. All models include SuperFrost technology which makes freezing simple and saves energy, it quickly reduces the temperature to −32 °C.
  • Electronic temperature control
  • All drawers can be removed to increase storage space