Desiccator cabinet, stacking, Dry-Keeper™, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H420530001
468-4111EA 11400 DKK
468-4111
Desiccator cabinet, stacking, Dry-Keeper™, SP Bel-Art
Ekssikatorer Ekssikatorkabinetter
This cabinet features perforated shelves that suit dry storage needs, cabinets can be stacked up to five units high.
  • Smoke-tinted, polystyrene
  • Two removable shelves can be set at four heights
Pull down door slides out of the way into a compartment located underneath the cabinet.
Leveringsinformation: Supplied with small desiccant tray.