Refrigerated/Heating Circulators, CORIO™ CP
Leverandør: JULABO GmbH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9013701 9013702 9013703 9013704 9013705 9013707 9013708 9013713.N1 9013713.S1 9013714.N1 9013714.S1 9013716.N1 9013715.N1 9013717.N1 9013728.N1
462-0630EA 37220 DKK
462-0630 462-0631 462-0632 462-0633 462-0634 462-0636 462-0637 461-1447 JULA9013713.S1 461-1448 JULA9013714.S1 461-1454 JULA9013715.N1 JULA9013717.N1 JULA9013728.N1
Refrigerated/Heating Circulators, CORIO™ CP
Cirkulatorsystemer Åbne cirkulatorsystemer Termostatbade til opvarmning/køling
CORIO refrigerated/heating circulators distinguish themselves with a great price performance ratio. They are ideal for all standard tasks and routine work in laboratories and industry.
  • Precise temperature control
  • Bright, white, easy to read display
  • Very quiet and easy operation
  • Have a USB interface and pump that can change between internal and external circulation via a lever located directly below the display
  • Integrated cooling coil for counter-cooling gives more usable space in the bath tank
  • Refrigeration unit without side vents and removable ventilation