Chlortetracyclin, HCl, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Chlortetracycline HCI, Aureomycin, 7-Chlorotetracycline hydrochloride, (4S,4aS,5aS,6S,12aS)-7-Chloro-4-(dimethylamino)-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrochloride

M-Clarity™ quality level = MQ200

46133-250MG-R
46133-250MG-REA 499 DKK
46133-250MG-R
Chlortetracyclin, HCl, Supelco®
Chlortetracyclin, HCl
Formel: C₂₂H₂₃ClN₂O₈·HCl
Opbevaringstemperatur: Køleskab
MDL Number: MFCD26142699
CAS-nummer: 64-72-2
EINECS: 200-591-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c30ef045b991708e740efca5e0b16e85e50dad56&vid=73859a4c8df27869a311529e828d95ae0616b4fb&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR