Circulating chillers, Microcool range
Leverandør: LAUDA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
L001046 L001047 L001048 L001049 L001051 L001052 L001053 L001054 L001055 L001056 L001057 L001058 L001059 L001060
461-1328EA 23970 DKK
461-1328 461-1329 461-1331 461-1332 LAUDL001051 LAUDL001052 LAUDL001053 LAUDL001054 LAUDL001055 LAUDL001056 LAUDL001057 LAUDL001058 LAUDL001059 LAUDL001060
Circulating chillers, Microcool range
Cirkulatorsystemer Lukkede cirkulatorsystemer Køletermostatbade
The Microcool range of circulation chillers includes five compact models providing cooling capacities from 0,25 to 1,2 kW with working temperatures from −10 up to +40 °C. An RS232 interface and alarm contact are integrated as standard, enabling system integration into processes without additional costs.
  • User interface with large LED display and a membrane keyboard make the units easy to use
  • High-quality block pump with magnetic coupling. The magnetic coupling of pump and electric motor avoid sealing problems on the pump shaft
  • Chillers have an autostart