Supelco® Organic Reference Standard, Geranyl acetate

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

45896-1ML-F
45896-1ML-FEA 325 DKK
45896-1ML-F
Supelco® Organic Reference Standard, Geranyl acetate
Standarder Økologiske standarder

  • GHS signal: GHS07


Advarsel: P261 - P264 - P272 - P273 - P280 - P302 + P352

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21c3f996a6b334b1c34f63e43ef1c3d1707f0657&vid=21c3f996a6b334b1c34f63e43ef1c3d1707f0657&prid=web&clid=SAPDAM

Bestil nu

Specifikationer

Synonym(s) trans-3,7-Dimethyl-2,6-octadien-1-yl acetate; trans-3,7-Dimethyl-2,6-octadienyl acetate
Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 105-87-3
MDL No. MFCD00015037
Formel CH₃CO₂CH₂CH=C(CH₃)CH₂CH₂CH=C(CH₃)₂
Molecular weight 196,29
Kogepunkt 138 °C/25 mmHg (lit.)
Fareklassebeskrivelse H315 - H317 - H412
Flammepunkt 104 °C - Closed cup
Shipping conditions STD
Storage conditions 2…8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR