Silicium 10.000 mg/l i vand med spor af flussyre (from (NH4)2SiF6) ARISTAR® enkeltelement standard til ICP
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
456012YEA 2360 DKK
456012Y
Silicium 10.000 mg/l i vand med spor af flussyre (from (NH4)2SiF6) ARISTAR® enkeltelement standard til ICP
Standarder Kationiske standarder Silicium Standards
(NH₄)₂ SiF₆ in H₂O tr. HF.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.