Rubidium 10.000 mg/l i fortyndet salpetersyre (from RbNO3) ARISTAR® enkeltelement standard til ICP
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
455912YEA 3300 DKK
455912Y
Rubidium 10.000 mg/l i fortyndet salpetersyre (from RbNO3) ARISTAR® enkeltelement standard til ICP
Standarder Kationiske standarder Rubidium Standards
RbNO₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.