Molybdænstandard, 1g/l Mo 1% ammoniumhydroxid (from (NH4)2MoO4) ARISTAR® enkeltelement standard til ICP
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
455682NEA 684 DKK
455682N 455684P
Molybdænstandard, 1g/l Mo 1% ammoniumhydroxid (from (NH4)2MoO4) ARISTAR® enkeltelement standard til ICP
Standarder Kationiske standarder Molybdenum Standards
(NH₄)₂MoO₄ in water tr. NH₄OH.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.