tri-Natriumcitrat, anhydrous 99% (by titrimetric analysis)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Sodium Citrate, Sodium citrate tribasic, Citric acid trisodium salt, Sodium 2-hydroxypropane-1,2,3-tricarboxylate

45556.A1 45556.30 45556.A7
45556.A1EA 1480 DKK
45556.A1 45556.30 45556.A7
tri-Natriumcitrat, anhydrous 99% (by titrimetric analysis)
tri-Natriumcitrat

Trisodium citrate is used in food additives, beverages, instant drinks, pharmaceuticals and detergents. It is also used as buffering agent, acidity regulator, sequestering or emulsifying agent.


It is an effective agent for removal of carbonate scale from boilers and for cleaning automobile radiators. In the medical field, it is used as an osmotic laxative and an active component of the WHO oral rehydration solution. It is also employed to control acidity in some substances such as gelatin desserts. Further, it is used as a flavour or as a preservative. In addition to this, it is used for the preservation of blood in blood banks.

Formel: C₆H₅Na₃O₇
Molekylvægt: 258.07 g/mol
Smeltepunkt: 300 °C
Massefylde: 1.815 g/cm³ (20 °C)
Flammepunkt: 290
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00012462
CAS-nummer: 68-04-2
EINECS: 200-675-3
Merck Index: 14,8602

Bestil nu

Specifikationer Testresultater

Assay (Titration) >98.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR