Thallium standard dissolved in oil, 1,000 µg/g Tl in mineral oil Specpure® for wear metal analysis

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

45306.18
45306.18EA 1140 DKK
45306.18
Thallium standard dissolved in oil, 1,000 µg/g Tl in mineral oil Specpure® for wear metal analysis
Standarder Kationiske standarder Thallium Standards

Physical Form: Liquid
MDL No.: MFCD01095325

Bestil nu

Specifikationer

Environmentally Preferable
MDL No. MFCD01095325

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR