Bunsen burner, cartridge type
Leverandør: USBECK Laborgeräte
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1422
451-0156EA 601 DKK
451-0156
Bunsen burner, cartridge type
Varmeapparater Gasbrændere
Nickel plated brass Bunsen burner ideal for heating or sterilising instruments and laboratory glassware.
  • With air regulation needle valve
Height: 85 mm
Bestillingsoplysninger: Supplied without cartridge, please order cartridge separately.