Jern(III)oxid, powder, NanoArc® 20-40nm APS, S.A. 30-60m2/g for cosmetic

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Ferric oxide, Ferric Oxide, (iron(III) oxide), Diiron trioxide

44895.22 44895.36 44895.14
44895.22EA 1540 DKK
44895.22 44895.36 44895.14
Jern(III)oxid, powder, NanoArc® 20-40nm APS, S.A. 30-60m2/g for cosmetic
Jern(III)oxid

NanoArc® cosmetic iron oxide is an auburn material offering unique cosmetic qualitites. The small particle size results in a strong Colour development combined with high translucency for a unique glowing look.


CAS No.: 1309-37-1 Molecular Formula: Fe2O3 Formula Weight: 159.69 Physical Form: 20-40nm APS Powder, S.A. 30-60m2/g Density: 5.24 Appearance: Red-brown Odour: Odourless Melting Point: 1565° MDL No.: MFCD00011008


NanoArc® cosmetic iron oxide is an auburn material offering unique cosmetic qualitites. The small particle size results in a strong Colour development combined with high translucency for a unique glowing look

Formel: Fe₂O₃
Smeltepunkt: 1565 °C
Massefylde: 5,24
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011008
CAS-nummer: 1309-37-1
EINECS: 215-168-2
Merck Index: 13,04058

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR