Praseodym(III,IV)oxid ≥99% (REO, rare earth oxide basis)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

44594.30 44594.A1
44594.30EA 1700 DKK
44594.30 44594.A1
Praseodym(III,IV)oxid ≥99% (REO, rare earth oxide basis)
Praseodymium(III,IV) oxide

Purity: 99% CAS No.: 12037-29-5 Molecular Formula: Pr6O11 Formula Weight: 1021.43 Physical Form: Powder Melting Point: 2500° MDL No.: MFCD00011178

Formel: O₁₁Pr₆
Molekylvægt: 1021.44 g/mol
Smeltepunkt: 2500 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011178
CAS-nummer: 12037-29-5
EINECS: 234-857-9

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR