Vand, Electran Molecular_Biology

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Hydrogen oxide

445847DEA 1170 DKK
445847D
Vand, Electran Molecular_Biology
Vand
Formel: H₂O
Molekylvægt: 18.02 g/mol
Kogepunkt: 100
Smeltepunkt: 0
Massefylde: 1 g/cm³ (4 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011332
CAS-nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Bestil nu

Specifikationer Testresultater

Osmolality (mOsm/kg) Max. 3
DNases Not detected
RNases Not detected
Proteases Not detected
Fe (Iron) Max. 0.006 ppm
Mg (Magnesium) Max. 0.005 ppm
Pb (Lead) Max. 0.005 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR