Bottle/tube rollers
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
88881004
444-0960EA 9720 DKK
444-0960
Bottle/tube rollers
Rystere Roterende og vuggende rysteborde
This sleek compact design unit is capable of rolling multiple size tubes or bottles for applications like cell culture, hybridisation assays and liquid mixing. The anti-corrosive, wear-resistant construction enables use in humid and CO₂ environments.
  • Adjustable speed accommodates a variety of applications
  • Adjustable rollers accommodate a variety of tube/bottle sizes
  • Stacking kit allows two units to be stacked, saving valuable bench space
  • Optional remote control facilitates operating unit while in incubator and adds timer function
Certificeringer: More Product Information