Kieselgel, granules -6+16 mesh desiccant, with indicator

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

44389.30 44389.A1 44389.A7
44389.30EA 575 DKK
44389.30 44389.A1 44389.A7
Kieselgel, granules -6+16 mesh desiccant, with indicator
Silica gel

Re-use after being saturated with water by heating to 121 °C for approximately 8 hours


CAS No.: 7631-86-9 Physical Form: Granules Sensitivity: Hygroscopic Appearance: Blue to red Odour: Odourless pH: 6 MDL No.: MFCD00011232

Formel: SiO₂
Molekylvægt: 60.08 g/mol
Kogepunkt: 2230
Smeltepunkt: 1710
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011232
CAS-nummer: 63231-67-4
EINECS: 231-545-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR