3,6-Dichloro-1,2-benzenedithiol 95%, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

442828-1G
442828-1GEA 2040 DKK
442828-1G
3,6-Dichloro-1,2-benzenedithiol 95%, Sigma-Aldrich®
3,6-Dichloro-1,2-benzenedithiol

3,6-Dichloro-1,2-benzenedithiol has been used as a ligand in thiolate complexes, and a series of homochalcogenide and mixed-chalcogenide ligand complexes.

Formel: Cl₂C₆H₂(SH)₂
Molekylvægt: 211.13 g/mol
Smeltepunkt: 58...60 °C (lit.)
Opbevaringstemperatur: Room Temperature
MDL Number: MFCD00216665
CAS-nummer: 87314-49-6
UN nummer: 2811
ADR: 6.1 II

Bestil nu

Specifikationer Testresultater

Appearance (Colour) Off White to Green to Beige and Faint Orange to Orange
Appearance (Form) Powder or Crystal or Chunk(s)
Infrared spectrum Conforms to Structure
Proton NMR spectrum Conforms to Structure
Titration by AgNO3, After Oxygen Combustion 94.0 - 106.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR