StarFish multiple experiment workstation
Leverandør: Heidolph Instruments GmbH & Co.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
505-80020-00 505-80025-00 505-80026-00 505-80040-00 505-80042-00 505-81010-00 505-81050-00 505-81060-00 505-81000-00 505-81001-00 505-80001-00
442-0614EA 2230 DKK
442-0614 442-0619 442-0620 442-0624 442-0626 442-0645 442-0649 442-0650 442-0643 442-0644 442-0604
StarFish multiple experiment workstation
Omrørere Magnetomrørere med varme
Converts a magnetic stirrer into a multiple experiment workstation, improve the productivity of your laboratory; use a single stirring hotplate to do multiple experiments. Perfect for simple heating/stirring experiments or more complex applications such as synthesis, concentrations, extractions and distillations. Accepts a wide range of standard laboratory glassware including vials, test tubes, beakers and round bottom flasks, a range of inserts are available for small vials up to 250 ml round bottom flasks.
  • Powerful stirring and heating of all positions simultaneously
  • Truly versatile,