VWR® VOS 40 digital & VOS 60 control, Electronic overhead stirrers
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
441-0304EA 8300 DKK
441-0304 441-0302 441-0303 441-0305 441-0306 441-0307
VWR® VOS 40 digital & VOS 60 control, Electronic overhead stirrers
Omrørere Overhead omrørere
Laboratory stirrers that are ideal for simple stirring tasks, suitable for quantities up to 25 L (VOS 40 digital), or for intensive stirring tasks for quantities up to 40 L (VOS 60 control). Due to microprocessor-controlled technology, the stirrers automatically adjust the speed within the speed range of 0/30 to 2000 min⁻¹.
  • Digital speed display
  • Infinitely adjustable speed
  • Push-through agitator shafts
  • Overload protection and units can be operated for short-term periods in overload conditions
  • Error code display
Safety circuits ensureMore Product Information