Zinksulfat heptahydrat 99.0-103.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Leverandør: Avantor
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

4382-01EA 701 DKK
4382-01 4382-R
Zinksulfat heptahydrat 99.0-103.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Zinksulfat heptahydrat
Formel: ZnSO₄·7H₂O
Molekylvægt: 287.54 g/mol
Smeltepunkt: 100
Massefylde: 1.97 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149894
CAS-nummer: 7446-20-0
EINECS: 231-793-3
UN nummer: 3077
ADR: 9 III
Merck Index: 12,10293

Bestil nu

Specifikationer Testresultater

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (ZnSO₄·7H₂O) 99.0 - 103.0 %
ACS - Insoluble Matter ≤0.005 %
ACS - pH of 5% Solution at 25°C 4.4 - 6.0
ACS - Nitrate (NO₃) ≤0.002 %
ACS - Ammonium (NH₄) ≤0.001 %
ACS - Calcium (Ca) ≤0.005 %
ACS - Lead (Pb) ≤0.003 %
ACS - Magnesium (Mg) ≤0.005 %
ACS - Potassium (K) ≤0.01 %
ACS - Sodium (Na) ≤0.05 %
Chloride (Cl) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Trace Impurities - Manganese (Mn) ≤2.000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR