(±)-Dihydrolipoic acid, Millipore®

Leverandør: Merck Millipore Calbiochem
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

437694-50
437694-50EA 1140 DKK
437694-50
(±)-Dihydrolipoic acid, Millipore®
(±)-Dihydrolipoic acid
Formel: C₈H₁₆O₂S₂
MDL Number: MFCD00166974
CAS-nummer: 7516-48-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR