Iridium ≥99.9% (metals basis), wire, Ø 0.125 mm (0.005 in)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

43208.BSEA 3540 DKK
43208.BS 43208.BW
Iridium ≥99.9% (metals basis), wire, Ø 0.125 mm (0.005 in)
Iridium

Purity: 99.9% CAS No.: 7439-88-5 Physical Form: ≈0.28g/m MDL No.: MFCD00011062

Formel: Ir
Kogepunkt: 5300 °C (1013 hPa)
Smeltepunkt: 2450 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011062
CAS-nummer: 7439-88-5
EINECS: 231-095-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR