Tetrabutylammoniumchlorid, hydrat 96%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Tetrabutyl ammonium chloride xhydrate, Tetra-n-butylammonium chloride hydrate, Tetrabutylammonium chloride hydrate, TBAC

432060050 432060250 432061000
432060050.EA 229 DKK
432060050. 432060250. 432061000.
Tetrabutylammoniumchlorid, hydrat 96%
Tetrabutylammoniumchlorid, hydrat
Formel: C₁₆H₃₆ClN·xH₂O
Smeltepunkt: 37…39 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011635
CAS-nummer: 37451-68-6

Bestil nu

Specifikationer Testresultater

Appearance Off-white Low melting solid
Infrared spectrum Conforms
Titration Argentometric ≥95.0 % (On dried substance)
Water ≤10.0 % (K.F.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR