Boss Head
Leverandør: Heidolph Instruments GmbH & Co.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
570-31100-00
431-0043EA 324 DKK
431-0043
Boss Head
Klemmer Krydsmuffe
Boss head made of anodised aluminum, used to fix retort clamps to stand rods for perfect vertical alignment.
Suitable for stand rods with up to 13,1 mm maximum diameter.