Palladium(II)acetat trimer (≥47% Pd) ≥99.98% (metals basis)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Palladium diacetate, Palladium (II) acetate

43086.03 43086.06
43086.03EA 2240 DKK
43086.03 43086.06
Palladium(II)acetat trimer (≥47% Pd) ≥99.98% (metals basis)
Palladium(II)acetat

Purity: 99.98% CAS No.: 3375-31-3 Molecular Formula: C12H18O12Pd3 Formula Weight: 673.46 Physical Form: Powder Appearance: Red-brown Melting Point: ca 205° dec. Merck Reference: 14,6991 MDL No.: MFCD00012453

Formel: C₁₂H₁₈O₁₂Pd₃
Molekylvægt: 673.46 g/mol
Smeltepunkt: ∼ 205 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00012453
CAS-nummer: 3375-31-3
EINECS: 222-164-4
Merck Index: 14,6991

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR