Butylacetat, dry_extra 99+%, AcroSeal®

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Butyl ethanoate, Acetic acid butyl ester, Acetic acid n-butyl ester, n-Butyl acetate

429230010 429231000
429230010.EA 1610 DKK
429230010. 429231000.
Butylacetat, dry_extra 99+%, AcroSeal®
Butylacetat
Formel: CH₃COO(CH₂)₃CH₃
Molekylvægt: 116.16 g/mol
Kogepunkt: 127 °C (760 mmHg)
Smeltepunkt: –78 °C
Massefylde: 0.88 g/cm³
Flammepunkt: 27 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009445
CAS-nummer: 123-86-4
UN nummer: 1123
ADR: 3 III
Merck Index: 13,01534
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=403329de943f8cb2ea6cd9d73ee35957d2140aa7&vid=682ab82de177b2985e94089408f7efebde59926d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.0 %

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR