Hydrogenchlorid ∼0.5 M methanol, AcroSeal®

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

428931000
428931000.EA 698 DKK
428931000.
Hydrogenchlorid ∼0.5 M methanol, AcroSeal®
Hydrogen chloride
Formel: HCl
Molekylvægt: 36.46 g/mol
Massefylde: 0.8 g/cm³
Flammepunkt: 12 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00011324
CAS-nummer: 7647-01-0
UN nummer: 3286
ADR: 3 II

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to light yellow Solution
Strength 0.4 to 0.6 M
Melting point 143 to 149 °C
Titration Argentometric ≥94 %

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR