Organic reference standard, all-trans-Astaxanthin, analytical standard, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

41659-1MG
41659-1MGEA 2190 DKK
41659-1MG
Organic reference standard, all-trans-Astaxanthin, analytical standard, Supelco®
Standarder Økologiske standarder

Astaxanthin is a potent carotenoid antioxidant found in marine algae, red yeast and many other plant and animal sources. Animal studies indicate that it reduces blood glucose and ameliorates several parameters of the diabetic metabolic syndrome.


It improves blood flow and vascular tone in models of hypertension. In vitro studies indicate that it upregulates connexin 43 and thus, may be chemopreventive against cancer.


Emballering: Bottomless glass bottle contents are inside inserted with fused cone.

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d602cb5efc65b9602c12890303aee2f34406a32&vid=6d602cb5efc65b9602c12890303aee2f34406a32&prid=web&clid=SAPDAM

Bestil nu

Specifikationer

Beskrivelse all-trans-Astaxanthin
Environmentally Preferable
CAS-nr 472-61-7
Formel C₄₀H₅₂O₄

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR