Syringeless filters, Claristep®
Leverandør: Sartorius
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
17C07FT-96 17C06FT-480 17C06FT-96 17C07FT-480
415-0000EA 2180 DKK
415-0000 415-0003 415-0002 415-0001
Syringeless filters, Claristep®
Filtre Filtre uden sprøjter
Claristep® filter units with PP housing and RC membranes are used for sample preparation by particle removal prior to analytics. The filters are suitable for small sample volumes of 60 to 600 µl.
  • Effective filtration diameter of 9,7 mm
  • Optimised for filtration of aqueous liquids as well as harsh solvents
  • Maximum chemical compatibility
  • Exceptionally low non-specific binding of analytes
  • Hold up volume <30 µl
  • Units can be sterilised by autoclaving or EO
The Claristep® station enables the processing of Claristep® filters. Simply position the sampleMore Product Information