Glassy carbon, spherical powder 630-1000 µm Type 1

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

41497.18 41497.30
41497.18EA 2340 DKK
41497.18 41497.30
Glassy carbon, spherical powder 630-1000 µm Type 1
Glassy carbon
Formel: C
Molekylvægt: 12.01 g/mol
Smeltepunkt: 3652
Massefylde: 1.95 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00133992
CAS-nummer: 7440-44-0
EINECS: 231-153-3
UN nummer: 1325
ADR: 4.1 III
Merck Index: 13,01818

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR