"Formalingskrukker ""comfort"" til PM 100, PM 100CM, PM 200 og PM 400"
Leverandør: Retsch GmbH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
01.462.0239 01.462.0240
412-0083EA 5880 DKK
412-0083 412-0084
"Formalingskrukker ""comfort"" til PM 100, PM 100CM, PM 200 og PM 400"
Dåser Grinding Jars/Bowls
Effective grinding can be achieved by the selection of grinding jars and grinding balls from a wide range of sizes and materials of construction. The ‘comfort’ grinding jars have several safety and design features that make them easy to use.
  • O-ring for gas-tight and dust-proof seal
  • Stainless steel protective jacket for jars that are not made of steel
  • Gap between flanges of jar and edge of lid for easy opening